Assay Details
| Gene Symbol: AT1G25530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102364.4 | NP_173924.1 | 4 - 5 | 1229 | 76 | |
| GenBank mRNA | AK226595.1 | - | 4 - 5 | 1095 | 76 | |
| BT022015.1 | - | 4 - 5 | 1095 | 76 | ||
More Information
Set Membership: |
|

AT1G25530
Genomic Map