Assay Details
| Gene Symbol: PFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084127.1 | NP_001077596.1 | 10 - 11 | 1955 | 81 | |
| NM_102365.5 | NP_173925.3 | 11 - 12 | 2141 | 81 | ||
| GenBank mRNA | AY170377.1 | - | 11 - 12 | 1772 | 81 | |
More Information
Set Membership: |
|

PFT1
Genomic Map