Assay Details
| Gene Symbol: PFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084127.1 | NP_001077596.1 | 12 - 13 | 2105 | 65 | |
| NM_102365.5 | NP_173925.3 | 13 - 14 | 2291 | 65 | ||
| GenBank mRNA | AY170377.1 | - | 13 - 14 | 1922 | 65 | |
More Information
Set Membership: |
|

PFT1
Genomic Map