Assay Details
| Gene Symbol: PFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084127.1 | NP_001077596.1 | 2 - 3 | 383 | 87 | |
| NM_102365.5 | NP_173925.3 | 3 - 4 | 569 | 87 | ||
| GenBank mRNA | AY170377.1 | - | 3 - 4 | 200 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001084127.1 transcripts.
Set Membership: |
|

PFT1
Genomic Map