Assay Details
| Gene Symbol: PFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084127.1 | NP_001077596.1 | 4 - 5 | 714 | 71 | |
| NM_102365.5 | NP_173925.3 | 5 - 6 | 900 | 71 | ||
| GenBank mRNA | AY170377.1 | - | 5 - 6 | 531 | 71 | |
More Information
Set Membership: |
|

PFT1
Genomic Map