Assay Details
| Gene Symbol: AT1G26170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102382.3 | NP_173942.2 | 6 - 7 | 906 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G26170| Gene Symbol: AT1G26170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102382.3 | NP_173942.2 | 6 - 7 | 906 | 97 | |
Set Membership: |
|