Assay Details
| Gene Symbol: AT1G26370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102401.4 | NP_173961.3 | 10 - 11 | 2043 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G26370| Gene Symbol: AT1G26370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102401.4 | NP_173961.3 | 10 - 11 | 2043 | 94 | |
Set Membership: |
|