Assay Details
| Gene Symbol: AT1G26370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102401.4 | NP_173961.3 | 6 - 7 | 1264 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G26370| Gene Symbol: AT1G26370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102401.4 | NP_173961.3 | 6 - 7 | 1264 | 137 | |
Set Membership: |
|