Assay Details
| Gene Symbol: AT1G26650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102427.4 | NP_564252.2 | 1 | 1382 | 172 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G26650| Gene Symbol: AT1G26650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102427.4 | NP_564252.2 | 1 | 1382 | 172 | |
Set Membership: |
|