Assay Details
| Gene Symbol: GALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332728.1 | NP_001319087.1 | 3 - 4 | 1132 | 110 | |
| NM_102445.4 | NP_174003.1 | 2 - 3 | 1092 | 110 | ||
| GenBank mRNA | AY133724.1 | - | 2 - 3 | 1072 | 110 | |
| EF428439.1 | - | 1 - 2 | 785 | 110 | ||
| KJ138985.1 | - | 1 - 2 | 785 | 110 | ||
More Information
Set Membership: |
|

GALT1
Genomic Map