Assay Details
| Gene Symbol: GALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332728.1 | NP_001319087.1 | 8 - 9 | 2059 | 71 | |
| NM_102445.4 | NP_174003.1 | 7 - 8 | 2019 | 71 | ||
| GenBank mRNA | AY133724.1 | - | 7 - 8 | 1999 | 71 | |
| EF428439.1 | - | 6 - 7 | 1712 | 71 | ||
| KJ138985.1 | - | 6 - 7 | 1712 | 71 | ||
More Information
Set Membership: |
|

GALT1
Genomic Map