Assay Details
| Gene Symbol: AT1G27060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102468.2 | NP_174026.1 | 3 - 4 | 628 | 65 | |
| GenBank mRNA | BT030023.1 | - | 3 - 4 | 512 | 65 | |
More Information
Set Membership: |
|

AT1G27060
Genomic Map