Assay Details
| Gene Symbol: AT1G27150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102477.4 | NP_564271.1 | 11 - 12 | 1341 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G27150| Gene Symbol: AT1G27150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102477.4 | NP_564271.1 | 11 - 12 | 1341 | 81 | |
Set Membership: |
|