Assay Details
| Gene Symbol: AT1G27210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102483.3 | NP_564273.1 | 1 - 2 | 1114 | 87 | |
| GenBank mRNA | AY057540.1 | - | 1 - 2 | 964 | 87 | |
| AY140087.1 | - | 1 - 2 | 964 | 87 | ||
| BT008391.1 | - | 1 - 2 | 855 | 87 | ||
More Information
Set Membership: |
|

AT1G27210
Genomic Map