Assay Details
| Gene Symbol: NTF2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102493.3 | NP_174051.1 | 1 - 2 | 507 | 76 | |
| GenBank mRNA | AY042889.1 | - | 1 - 2 | 401 | 76 | |
| AY072473.1 | - | 1 - 2 | 295 | 76 | ||
More Information
Set Membership: |
|

NTF2A
Genomic Map