Assay Details
| Gene Symbol: TOM20-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332759.1 | NP_001322710.1 | 5 - 6 | 752 | 148 | |
| NM_102502.3 | NP_174059.2 | 5 - 6 | 640 | 148 | ||
| GenBank mRNA | AK228213.1 | - | 5 - 6 | 591 | 148 | |
| BT024602.1 | - | 5 - 6 | 502 | 148 | ||
More Information
Set Membership: |
|

TOM20-2
Genomic Map