Assay Details
| Gene Symbol: EX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102515.3 | NP_564287.1 | 8 - 9 | 1852 | 130 | |
| GenBank mRNA | AY045812.1 | - | 8 - 9 | 1754 | 130 | |
| AY114085.1 | - | 8 - 9 | 1684 | 130 | ||
More Information
Set Membership: |
|

EX2
Genomic Map