Assay Details
| Gene Symbol: APL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102533.5 | NP_174089.1 | 2 - 3 | 771 | 105 | |
| GenBank mRNA | AY063927.1 | - | 2 - 3 | 625 | 105 | |
| AY091251.1 | - | 2 - 3 | 331 | 105 | ||
More Information
Set Membership: |
|

APL2
Genomic Map