Assay Details
| Gene Symbol: APL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102533.5 | NP_174089.1 | 5 - 6 | 1113 | 130 | |
| GenBank mRNA | AY063927.1 | - | 5 - 6 | 967 | 130 | |
| AY091251.1 | - | 5 - 6 | 673 | 130 | ||
More Information
Set Membership: |
|

APL2
Genomic Map