Assay Details
| Gene Symbol: DPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102563.4 | NP_174119.1 | 5 - 6 | 997 | 122 | |
| GenBank mRNA | AB175035.1 | - | 5 - 6 | 691 | 122 | |
| AF360166.1 | - | 5 - 6 | 750 | 122 | ||
| AY113914.1 | - | 5 - 6 | 691 | 122 | ||
More Information
Set Membership: |
|

DPL1
Genomic Map