Assay Details
| Gene Symbol: DPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102563.4 | NP_174119.1 | 9 - 10 | 1415 | 97 | |
| GenBank mRNA | AB175035.1 | - | 9 - 10 | 1109 | 97 | |
| AF360166.1 | - | 9 - 10 | 1168 | 97 | ||
| AY113914.1 | - | 9 - 10 | 1109 | 97 | ||
More Information
Set Membership: |
|

DPL1
Genomic Map