Assay Details
| Gene Symbol: FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332805.1 | NP_001322847.1 | 1 - 2 | 372 | 73 | |
| NM_102585.3 | NP_174141.1 | 1 - 2 | 358 | 73 | ||
| GenBank mRNA | AF174428.2 | - | 1 - 2 | 232 | 73 | |
| AF370177.1 | - | 1 - 2 | 289 | 73 | ||
| AY056389.1 | - | 1 - 2 | 255 | 73 | ||
| AY059137.1 | - | 1 - 2 | 223 | 73 | ||
More Information
Set Membership: |
|

FIP1
Genomic Map