Assay Details
| Gene Symbol: FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332805.1 | NP_001322847.1 | 2 - 3 | 478 | 76 | |
| NM_102585.3 | NP_174141.1 | 2 - 3 | 464 | 76 | ||
| GenBank mRNA | AF370177.1 | - | 2 - 3 | 395 | 76 | |
| AY056389.1 | - | 2 - 3 | 361 | 76 | ||
| AY059137.1 | - | 2 - 3 | 329 | 76 | ||
| AY086994.1 | - | 2 - 3 | 361 | 76 | ||
More Information
Set Membership: |
|

FIP1
Genomic Map