Assay Details
| Gene Symbol: FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332805.1 | NP_001322847.1 | 3 - 4 | 747 | 89 | |
| NM_102585.3 | NP_174141.1 | 3 - 4 | 733 | 89 | ||
| GenBank mRNA | AF174428.2 | - | 3 - 4 | 607 | 89 | |
| AF370177.1 | - | 3 - 4 | 664 | 89 | ||
| AY056389.1 | - | 3 - 4 | 630 | 89 | ||
| AY059137.1 | - | 3 - 4 | 598 | 89 | ||
| AY086994.1 | - | 3 - 4 | 630 | 89 | ||
More Information
Set Membership: |
|

FIP1
Genomic Map