Assay Details
| Gene Symbol: ERF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102602.6 | NP_174158.1 | 1 | 926 | 160 | |
| GenBank mRNA | AY099798.1 | - | 1 | 841 | 161 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERF12| Gene Symbol: ERF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102602.6 | NP_174158.1 | 1 | 926 | 160 | |
| GenBank mRNA | AY099798.1 | - | 1 | 841 | 161 | |
Set Membership: |
|