Assay Details
| Gene Symbol: AT5G27280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122610.3 | NP_198080.1 | 2 - 3 | 321 | 65 | |
| GenBank mRNA | AY059931.1 | - | 2 - 3 | 258 | 65 | |
| AY081569.1 | - | 2 - 3 | 213 | 65 | ||
More Information
Set Membership: |
|

AT5G27280
Genomic Map