Assay Details
| Gene Symbol: HB-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102610.4 | NP_174164.2 | 16 - 17 | 4559 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HB-1| Gene Symbol: HB-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102610.4 | NP_174164.2 | 16 - 17 | 4559 | 150 | |
Set Membership: |
|