Assay Details
| Gene Symbol: HSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102612.4 | NP_174166.1 | 1 - 2 | 2788 | 73 | |
| GenBank mRNA | AK227198.1 | - | 1 - 2 | 2652 | 73 | |
| AY093235.1 | - | 1 - 2 | 2677 | 73 | ||
| BT008808.1 | - | 1 - 2 | 2587 | 73 | ||
| FJ708639.1 | - | 1 - 2 | 2587 | 73 | ||
More Information
Set Membership: |
|

HSL1
Genomic Map