Assay Details
| Gene Symbol: SYP61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036025.5 | NP_001031102.2 | 7 - 8 | 703 | 76 | |
| NM_001332815.1 | NP_001321010.1 | 4 - 5 | 575 | 76 | ||
| NM_102617.4 | NP_564310.1 | 7 - 8 | 821 | 76 | ||
| GenBank mRNA | AF355754.2 | - | 6 - 7 | 625 | 76 | |
| AY150420.1 | - | 6 - 7 | 625 | 76 | ||
More Information
Set Membership: |
|

SYP61
Genomic Map