Assay Details
| Gene Symbol: CIPK18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102663.2 | NP_174217.1 | 1 | 1340 | 86 | |
| GenBank mRNA | AY034099.1 | - | 1 | 1161 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CIPK18| Gene Symbol: CIPK18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102663.2 | NP_174217.1 | 1 | 1340 | 86 | |
| GenBank mRNA | AY034099.1 | - | 1 | 1161 | 86 | |
Set Membership: |
|