Assay Details
| Gene Symbol: UGE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334071.1 | NP_001321118.1 | 2 - 3 | 472 | 126 | |
| NM_104996.6 | NP_564811.1 | 2 - 3 | 521 | 126 | ||
| GenBank mRNA | AK117913.1 | - | 2 - 3 | 304 | 126 | |
| AY085887.1 | - | 2 - 3 | 449 | 126 | ||
| BT024882.1 | - | 2 - 3 | 253 | 126 | ||
More Information
Set Membership: |
|

UGE3
Genomic Map