Assay Details
| Gene Symbol: UGE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334071.1 | NP_001321118.1 | 6 - 7 | 1016 | 79 | |
| NM_104996.6 | NP_564811.1 | 6 - 7 | 1065 | 79 | ||
| GenBank mRNA | AK117913.1 | - | 6 - 7 | 848 | 79 | |
| AY085887.1 | - | 6 - 7 | 993 | 79 | ||
| BT024882.1 | - | 6 - 7 | 797 | 79 | ||
More Information
Set Membership: |
|

UGE3
Genomic Map