Assay Details
| Gene Symbol: CDKD1;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106028.4 | NP_177510.1 | 1 - 2 | 305 | 87 | |
| GenBank mRNA | AB047275.1 | - | 1 - 2 | 102 | 87 | |
| AY063843.1 | - | 1 - 2 | 267 | 87 | ||
| AY091227.1 | - | 1 - 2 | 102 | 87 | ||
More Information
Set Membership: |
|

CDKD1;1
Genomic Map