Assay Details
| Gene Symbol: ARP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334612.1 | NP_001323057.1 | 2 - 3 | 187 | 82 | |
| NM_001334613.1 | NP_001323058.1 | 2 - 3 | 187 | 82 | ||
| NM_106050.2 | NP_177531.1 | 2 - 3 | 187 | 82 | ||
| GenBank mRNA | BT029316.1 | - | 1 - 2 | 102 | 82 | |
More Information
Set Membership: |
|

ARP4A
Genomic Map