Assay Details
| Gene Symbol: ARP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334612.1 | NP_001323057.1 | 4 - 5 | 410 | 101 | |
| NM_106050.2 | NP_177531.1 | 4 - 5 | 410 | 101 | ||
| GenBank mRNA | BT029316.1 | - | 3 - 4 | 325 | 101 | |
More Information
Set Membership: |
|

ARP4A
Genomic Map