Assay Details
Gene Symbol: GR-RBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106083.5 | NP_177563.1 | 2 - 3 | 292 | 71 | |
GenBank mRNA | BT002037.1 | - | 2 - 3 | 188 | 71 | |
BT006315.1 | - | 1 - 2 | 119 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
