Assay Details
| Gene Symbol: AT5G27530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122635.2 | NP_198105.2 | 5 - 6 | 696 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G27530| Gene Symbol: AT5G27530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122635.2 | NP_198105.2 | 5 - 6 | 696 | 111 | |
Set Membership: |
|