Assay Details
| Gene Symbol: MIRO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036876.2 | NP_001031953.1 | 10 - 11 | 1567 | 85 | |
| NM_122636.4 | NP_198106.1 | 10 - 11 | 1526 | 85 | ||
| GenBank mRNA | AY081280.1 | - | 10 - 11 | 1518 | 85 | |
| BT003352.1 | - | 9 - 10 | 1354 | 85 | ||
More Information
Set Membership: |
|

MIRO1
Genomic Map