Assay Details
| Gene Symbol: MIRO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036876.2 | NP_001031953.1 | 3 - 4 | 560 | 81 | |
| NM_122636.4 | NP_198106.1 | 3 - 4 | 519 | 81 | ||
| GenBank mRNA | AY081280.1 | - | 3 - 4 | 511 | 81 | |
| BT003352.1 | - | 2 - 3 | 347 | 81 | ||
More Information
Set Membership: |
|

MIRO1
Genomic Map