Assay Details
| Gene Symbol: MIRO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036876.2 | NP_001031953.1 | 8 - 9 | 1277 | 105 | |
| NM_122636.4 | NP_198106.1 | 8 - 9 | 1236 | 105 | ||
| GenBank mRNA | AY081280.1 | - | 8 - 9 | 1228 | 105 | |
| BT003352.1 | - | 7 - 8 | 1064 | 105 | ||
More Information
Set Membership: |
|

MIRO1
Genomic Map