Assay Details
| Gene Symbol: AGL89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122640.2 | NP_198110.1 | 1 | 692 | 149 | |
| GenBank mRNA | AY141226.1 | - | 1 | 566 | 149 | |
| BT025291.1 | - | 1 | 566 | 149 | ||
| DQ446991.1 | - | 1 | 566 | 149 | ||
More Information
Set Membership: |
|

AGL89
Genomic Map