Assay Details
| Gene Symbol: AT5G27650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122647.3 | NP_198117.2 | 1 - 2 | 1253 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G27650| Gene Symbol: AT5G27650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122647.3 | NP_198117.2 | 1 - 2 | 1253 | 94 | |
Set Membership: |
|