Assay Details
| Gene Symbol: RECQSIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344029.1 | NP_001330794.1 | 16 - 17 | 3001 | 80 | |
| NM_001344030.1 | NP_001330793.1 | 16 - 17 | 2833 | 80 | ||
| NM_001344031.1 | NP_001330795.1 | 14 - 15 | 2838 | 80 | ||
| NM_122650.5 | NP_568499.1 | 15 - 16 | 3107 | 80 | ||
| GenBank mRNA | AJ404475.1 | - | 15 - 16 | 2648 | 80 | |
| AY059754.1 | - | 15 - 16 | 2919 | 80 | ||
More Information
Set Membership: |
|

RECQSIM
Genomic Map