Assay Details
| Gene Symbol: RECQSIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344027.1 | NP_001330792.1 | 4 - 5 | 1614 | 130 | |
| NM_001344028.1 | NP_001330796.1 | 5 - 6 | 1513 | 130 | ||
| NM_001344029.1 | NP_001330794.1 | 5 - 6 | 1508 | 130 | ||
| NM_001344030.1 | NP_001330793.1 | 5 - 6 | 1340 | 130 | ||
| NM_001344031.1 | NP_001330795.1 | 4 - 5 | 1426 | 130 | ||
| NM_122650.5 | NP_568499.1 | 4 - 5 | 1614 | 130 | ||
| GenBank mRNA | AJ404475.1 | - | 4 - 5 | 1155 | 130 | |
| AY059754.1 | - | 4 - 5 | 1426 | 130 | ||
| AY133823.1 | - | 4 - 5 | 1102 | 130 | ||
More Information
Set Membership: |
|

RECQSIM
Genomic Map