Assay Details
| Gene Symbol: RECQSIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344027.1 | NP_001330792.1 | 9 - 10 | 2306 | 63 | |
| NM_001344028.1 | NP_001330796.1 | 10 - 11 | 2205 | 63 | ||
| NM_001344029.1 | NP_001330794.1 | 10 - 11 | 2200 | 63 | ||
| NM_001344030.1 | NP_001330793.1 | 10 - 11 | 2032 | 63 | ||
| NM_001344031.1 | NP_001330795.1 | 8 - 9 | 2037 | 63 | ||
| NM_122650.5 | NP_568499.1 | 9 - 10 | 2306 | 63 | ||
| GenBank mRNA | AJ404475.1 | - | 9 - 10 | 1847 | 63 | |
| AY059754.1 | - | 9 - 10 | 2118 | 63 | ||
| AY133823.1 | - | 9 - 10 | 1794 | 63 | ||
More Information
Set Membership: |
|

RECQSIM
Genomic Map