Assay Details
| Gene Symbol: BGLU24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122734.2 | NP_198203.1 | 9 - 10 | 1191 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BGLU24| Gene Symbol: BGLU24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122734.2 | NP_198203.1 | 9 - 10 | 1191 | 96 | |
Set Membership: |
|