Assay Details
| Gene Symbol: FRS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344075.1 | NP_001331634.1 | 3 - 4 | 1550 | 65 | |
| NM_122736.2 | NP_198205.2 | 2 - 3 | 1473 | 65 | ||
More Information
Set Membership: |
|

FRS10
Genomic Map