Assay Details
| Gene Symbol: AN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122747.4 | NP_198216.2 | 1 - 2 | 529 | 101 | |
| GenBank mRNA | AY102639.1 | - | 1 - 2 | 123 | 101 | |
| BT002848.1 | - | 1 - 2 | 359 | 101 | ||
| BT004462.1 | - | 1 - 2 | 87 | 101 | ||
More Information
Set Membership: |
|

AN3
Genomic Map