Assay Details
| Gene Symbol: PAP26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122874.4 | NP_198334.1 | 4 - 5 | 1016 | 76 | |
| GenBank mRNA | AY050812.1 | - | 4 - 5 | 783 | 76 | |
| AY091415.1 | - | 3 - 4 | 710 | 76 | ||
More Information
Set Membership: |
|

PAP26
Genomic Map