Assay Details
| Gene Symbol: AT5G35320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122923.3 | NP_198382.1 | 1 - 2 | 252 | 59 | |
| GenBank mRNA | AY074341.1 | - | 1 - 2 | 152 | 59 | |
| AY117159.1 | - | 1 - 2 | 105 | 59 | ||
More Information
Set Membership: |
|

AT5G35320
Genomic Map